Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Open Head Pail 2.5 Gallon
SDP

Catalog No. 19120023
Change view
Click to view available options
Capacity (English):
2.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-120-023 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-023 Supplier Youngstown Barrel & Drum Supplier No. 2377NG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plastic Open Head Pail 2.5 Gallon w/screw lid. UN rated 1H2/Y19/S. White. Used for shipping and storing small quantities of hazardous and non-hazardous materials.

Store or ship smaller quantities of solids.

Specifications

Certifications/Compliance UN 1H2/Y19/S
Product Type Pail
Closure Type Screw Lid
Capacity (English) 2.5 gal.
Capacity (Metric) 9.46353 L
Color White
For Use With (Application) PACKAGING FOR HAZARDOUS SOLIDS
Material HDPE
Freight Class 250
Head Style Screw Lid
Includes Lid and Handle
Length (Metric) 25.908 cm
Width (Metric) 25.908 cm
Height (English) 10.44 in.
Height (Metric) 26.5176 cm
Diameter (English) Inner 10 in.
Diameter (English) Outer 10.20 in.
Diameter (Metric) Inner 25.4 cm
Diameter (Metric) Outer 25.908 mm
Weight (English) 2.09 lb.
Weight (Metric) 0.9480081 Kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.