Learn More
Uline Plastic Pail - 5 Gallon | Red | 1 EA
Catalog No. NC1260485
Encompass
Safely store or ship your products in these durable pails. High density polyethylene construction withstands temperatures up to 180F. Stackable with lid Empty pails nest for storage. FDA compliant. Lids sold separately. Optional Companion Box available.
Catalog No. NC1260485 Supplier Uline Supplier No. S7914R
Safely store or ship your products in these durable pails. High density polyethylene construction withstands temperatures up to 180F. Stackable with lid Empty pails nest for storage. FDA compliant. Lids sold separately. Optional Companion Box available.