Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials Plastic Pasteur Pipettes

Catalog No. p-200004261
Change view
Click to view available options
Length (Metric):
146 mm
230 mm
Material:
Plastic
Polystyrene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Material
03-391-280 146 mm Plastic
03-391-281 230 mm Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-391-280 Supplier Cole-Parmer™ Essentials Supplier No. 0439511
Only null left
Add to Cart
Add to Cart

A safe alternative to glass pipettes

  • Manufactured from medical-grade polystyrene in an ISO Class 8 cleanroom
  • Gamma irradiated, and RNase, DNase, and pyrogen free

Disposable polystyrene (PS) Pasteur pipettes offer the aspirating precision of glass, with the safety and benefits of plastic. Ideal for use with aspirating bulbs or vacuum applications to remove liquid waste from flasks, dishes, plates, and tubes. The long flexible stem can be bent to draw liquid from narrow or small volume tubes. Supplied individually wrapped.

Specifications

Material Plastic
Sterility Sterile
Graduated No
Length (English) 5.75 in.
Packaging Type Individually wrapped
Color Clear
Length (Metric) 146 mm
Type Pasteur Pipet
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.