Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Plastic pH Test Strips
SCI_ED

Catalog No. S04674 Shop All Fisherbrand Products
Change view
Click to view available options
pH Range:
0 to 14
0 to 6
5 to 9
5.5 to 8
6.5 to 13
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S04674 0 to 14
S04675 5.5 to 8
S04676 6.5 to 13
S04677 0 to 6
S04673 5 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S04674

missing translation for 'validMainframeMsg'

Offer excellent color separation in the pH range; 100 pH test strips/vial.

  • Packaged in flip-top, spill-proof vial
  • Colorimetric chart located on the vial label
  • Plastic pH test strip construction
  • Ideal for lab, field or classroom useInstant,
  • Instant dip and read result

Order Info

Box 6 Vials, 100 strips per vial.

Specifications

pH Range 0 to 14
For Use With (Application) For lab, field or classroom use, instant dip and read result
Graduations 1
Includes Colorimetric chart on the vial label
Quantity 100 Strips/Vial, 6 Vials/Pk.
Storage Requirements Room Temperature
Type Test Strip, PH PLAS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.