Learn More
Haztech Systems, Inc Hazcat Plastic Pipettes. 2ml Graduated
SDP
Catalog No. NC9249854
Encompass
Hazcat polyethylene transfer pipettes. These pipettes are the correct size to make gas collection bulbs when used with the Hazcat System. Graduated to 2ml. Bundle of 40ea.
Catalog No. NC9249854 Supplier Haztech Systems, Inc Supplier No. HW4049
Hazcat polyethylene transfer pipettes. These pipettes are the correct size to make gas collection bulbs when used with the Hazcat System. Graduated to 2ml. Bundle of 40ea.