Learn More
Foxx Life Sciences Borosil® Plastic Polypropylene Stopper for Volumetric Flasks with Interchangeable Joint 7/16, 100/CS
Catalog No. 501720464
Encompass_Preferred
Borosil® Plastic Polypropylene Flat Head Stoppers are with Interchangeable Joint and can be used with Volumetric Flask of 5645, 5647 families or for other suitable joints. Polypropylene Available in sizes of 7/16 to 34/35. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type I, Borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0464 Supplier Foxx Life Sciences Supplier No. 8300007
Borosil® Plastic Polypropylene Flat Head Stoppers are with Interchangeable Joint and can be used with Volumetric Flask of 5645, 5647 families or for other suitable joints. Polypropylene Available in sizes of 7/16 to 34/35. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type I, Borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.