Learn More
Sklar Instruments Plastic Procedure Tray, Sklar, 11-3/4 in. x 16 in. Plastic Procedure Tray Perforated. Reusable Plastic. Autoclavable. Features No-Slip Textured Surface. EA/1.
SDP
Catalog No. 501437162 Shop All Sklar Instruments Products
Encompass_Preferred
Plastic Procedure Tray, Sklar, 11-3/4 in. x 16 in. Plastic Procedure Tray Perforated. Reusable Plastic. Autoclavable. Features No-Slip Textured Surface.
This item is not returnable. View return policy
Catalog No. 50-143-7162 Supplier Sklar Instruments Supplier No. 101967
Plastic Procedure Tray, Sklar, 11-3/4 in. x 16 in. Plastic Procedure Tray Perforated. Reusable Plastic. Autoclavable. Features No-Slip Textured Surface.