Learn More
Bellco Glass, Inc. Plastic rack is manufactured from polypropylene and is autoclavable reusable and recyclable. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily and chemically resistant to drips. ¿Case Quantity 5
SDP
Catalog No. 502330593
Encompass_Preferred
Plastic rack is manufactured from polypropylene and is autoclavable reusable and recyclable. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily and chemically resistant to drips. ¿Case Quantity 5
Catalog No. 50-233-0593 Supplier Bellco Glass, Inc. Supplier No. 229429000
Plastic rack is manufactured from polypropylene and is autoclavable reusable and recyclable. Design of bottom prevents racks from sticking/freezing to surfaces. Widely spaced design catches drips and allows tubes to be retrieved more easily and chemically resistant to drips. ¿Case Quantity 5