Learn More
MSC Vikan Round Bowl Scoop, Polypropylene, 67.6 Fl oz, Yellow
Catalog No. 502836141
Encompass
This round bowl scoop is constructed from durable polypropylene and designed for optimal performance in the food industry. With a capacity of 67.6 fl oz and a vibrant yellow color, it is suitable for a wide range of applications from ingredient transfer to general use. The scoop is engineered to meet stringent international food contact regulations, ensuring safe and compliant operations.
  • Made from polypropylene for durability and hygiene.
  • 67.6 fl oz capacity for efficient material handling.
  • Yellow color for easy identification or HACCP compliance.
  • Complies with EU, FDA, and UK food contact regulations.
  • Suitable for aqueous, acidic, alcoholic, fatty, and dry food types.
  • Wide usage temperature range from -4 °F to 212 °F.
  • Recyclable product, supporting environmental initiatives.
  • Measures 13 in long, 7.7 in wide, and 4.5 in high.
Catalog No. 50-283-6141 Supplier MSC Supplier No. 39846746
This round bowl scoop is constructed from durable polypropylene and designed for optimal performance in the food industry. With a capacity of 67.6 fl oz and a vibrant yellow color, it is suitable for a wide range of applications from ingredient transfer to general use. The scoop is engineered to meet stringent international food contact regulations, ensuring safe and compliant operations.
  • Made from polypropylene for durability and hygiene.
  • 67.6 fl oz capacity for efficient material handling.
  • Yellow color for easy identification or HACCP compliance.
  • Complies with EU, FDA, and UK food contact regulations.
  • Suitable for aqueous, acidic, alcoholic, fatty, and dry food types.
  • Wide usage temperature range from -4 °F to 212 °F.
  • Recyclable product, supporting environmental initiatives.
  • Measures 13 in long, 7.7 in wide, and 4.5 in high.