Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Plastic Safety Can with Stainless Steel Piston Pump

Catalog No. 18666731 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-666-731 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-666-731 Supplier Justrite™ Supplier No. 14586
  • $1,344.00
    $864.65 / Each of 1

    Save $479.35

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All in one storage and dispensing solution for flammable or combustible liquids. This Type I, FM approved, safety can is fitted with a stainless steel pump that allows storing up to 5 gallons of flammable or combustible liquids while also allowing their dispensing at the point of use.

  • All in one storage and dispensing solution for flammable or combustible liquids.
  • This Type I, FM approved safety can is fitted with a stainless steel pump that allows storing up to 5 gallons of flammable or combustible liquids while also allowing their dispensing at the point of use.
  • The wide filling spout makes it easy to receive liquids from bulk containers and includes a self-closing cap that safely provides pressure relief in the event of a fire.
  • The attached manually operated piston pump delivers up to 3.5 oz. per stroke and a vacuum-relief valve allows liquids to flow smoothly in a controlled and safe manner.
  • The pump body is constructed with stainless steel with PTFE seals, offering superior chemical resistance and a long life.
  • Included internal flash arrestor keeps contents safe from external flame flashbacks.
  • Justrite's unique, conductive carbon insert within the container allows for proper grounding with only one connection point for proper static control.
  • Uniquely ideal for Class 1B and Class 1C alcohol based hand sanitizers.

Specifications

Capacity (English) 5 gal.
Capacity (Metric) 19 L
Color Red
For Use With (Application) Dispensing of Flammable Liquids
For Use With (Equipment) Flammable Liquids
Material Polyethylene (Can), Stainless Steel (Pump, Hardware)
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.