Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Sign Sliders Sign Board, 3-Side Free Stand, Plastic, 64H x 24W in.

Catalog No. 19111491 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
24 x 64 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-111-491 24 x 64 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-111-491 Supplier Brady™ Supplier No. 140738

Improve workplace safety with proper signage and markings. This Brady™ plastic, 3-sided sign board is a durable, clear and effective tool to raise awareness to workplace hazards.

Safety Sliders are visual communication platforms for your facility that will communicate your most important safety and awareness messages to facility visitors and employees.

Specifications

Material Plastic
Shape Rectangular
Dimensions (L x W) 24 x 64 in.
Displayed Text MANTENER LOS CILINDROS ENCADENADOS
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.