Learn More
StatLab Plastic Slide Jars and Caps
SDP
Catalog No. 22021374
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
22-021-374 Polypropylene
1 options
This item is not returnable. View return policy
Catalog No. 22-021-374 Supplier StatLab Supplier No. LWS24
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
StatLab Plastic Slide Jars and Caps
  • Stain up to four slides in only 20 mL to 40 mL of stain
  • Slide jar is leaktight, chemical resistant, and microwaveable
  • Perfect for routine or specialized staining procedures of four slides or less
  • Slide jar can be reused over and over again
  • 100/pack
Specifications
Autoclavable Not Autoclavable
Bottom Shape Flat
Color Natural
Disposable Reusable
Material Polypropylene
Sterility Non-sterile
Surface Treatment Non-treated
Quantity 100/Pk.