Learn More
Coopersurgical Inc Pap-Perfect Plastic Spatulas (500/Box)
Catalog No. NC1440943
Encompass_Preferred
Use the Medscand Pap-Perfect Plastic Spatula in combination with Cytobrush Plus or Cytobrush Plus GT. The spatula gently rubs the mucosa, ensuring a rich, complete sample from the ectocervix. Made of inert polypropylene plastic, the spatula will release collected cells more easily than a wooden spatula.
This item is not returnable. View return policy
Catalog No. NC1440943 Supplier Coopersurgical Inc Supplier No. 11080
Use the Medscand Pap-Perfect Plastic Spatula in combination with Cytobrush Plus or Cytobrush Plus GT. The spatula gently rubs the mucosa, ensuring a rich, complete sample from the ectocervix. Made of inert polypropylene plastic, the spatula will release collected cells more easily than a wooden spatula.