Learn More
MSC Vikan Shovel, Polypropylene, Lime, 37 1/2 in Length
Catalog No. 502839796
Encompass
This one-piece polypropylene food service shovel is designed for hygienic handling of bulk materials in food processing and other food handling applications. Its durable, lightweight construction, combined with a large blade and extended handle, provides optimal control for scooping.
  • Hygienic one-piece construction prevents harboring of dirt or germs.
  • Suitable for various uses including meat, dairy, seafood, and poultry processing.
  • Lime color can be used as part of an HACCP color-coded system for cross-contamination prevention.
  • Dishwasher safe for easy cleaning.
  • Autoclave safe for sterilization.
  • Made in America.
  • Color coded.
Catalog No. 50-283-9796 Supplier MSC Supplier No. 95111985
This one-piece polypropylene food service shovel is designed for hygienic handling of bulk materials in food processing and other food handling applications. Its durable, lightweight construction, combined with a large blade and extended handle, provides optimal control for scooping.
  • Hygienic one-piece construction prevents harboring of dirt or germs.
  • Suitable for various uses including meat, dairy, seafood, and poultry processing.
  • Lime color can be used as part of an HACCP color-coded system for cross-contamination prevention.
  • Dishwasher safe for easy cleaning.
  • Autoclave safe for sterilization.
  • Made in America.
  • Color coded.