Learn More
Cole-Parmer Cole-Parmer CP143 Plastic and SS Dissecting Blade Handle, #8 for #60 and #70 Blades; 1/Pk
Catalog No. NC1461593 Shop All Cole-Parmer Products
Encompass_Preferred
Choose from carbon steel or stainless steel scalpel blades. This blade handle can be used with dissecting blades: 10822-14, 10822-15.
Catalog No. NC1461593 Supplier Cole-Parmer Supplier No. 1082234
Choose from carbon steel or stainless steel scalpel blades. This blade handle can be used with dissecting blades: 10822-14, 10822-15.