Learn More
GLOMED INC Plastic Sterilization Tray 6.5'' x 2.5'' x 0.75''
Supplier:  GLOMED INC CP614
Encompass_Preferred
Size: 6'' x 2'' x 0.75'' (Internal Dimensions) Designed to protect delicate instruments during sterilization. Enhanced features include a slotted lid and raised node base surface to facilitate the circulation of steam and aid in drying. Silicone pin mats included for added protection. Made from the polymer Ultem® to ensure strength and long lasting durability. Transparent lid for easy identification of contents.Trays are stackable
Catalog No. NC1400735