Learn More
Grainger Plastic Strapping Hand Strapping 5/8
Catalog No. 502764204
Encompass_Preferred
The Grainger 16P069 plastic strapping is a 5/8" wide polypropylene hand strapping designed for light to medium duty bundling, bailing, and unitizing applications. It has a break strength of 250 lbs, is available in clear or colors in multiple roll lengths up to 1500 ft, and works with standard friction weld and seal joint tools for securing bundles. This versatile, economical strapping provides good break strength and UV resistance for indoor and protected outdoor use.
Catalog No. 50-276-4204 Supplier Grainger Supplier No. 16P069
The Grainger 16P069 plastic strapping is a 5/8" wide polypropylene hand strapping designed for light to medium duty bundling, bailing, and unitizing applications. It has a break strength of 250 lbs, is available in clear or colors in multiple roll lengths up to 1500 ft, and works with standard friction weld and seal joint tools for securing bundles. This versatile, economical strapping provides good break strength and UV resistance for indoor and protected outdoor use.