Learn More
Grainger Plastic Strapping Hand Strapping 3/4
Catalog No. 502764206
Encompass_Preferred
The Grainger 16P074 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is UL/CSA approved. The 16P074 is ideal for driving pumps, fans, blowers, conveyors, and other industrial machinery requiring up to 3/4 horsepower.
Catalog No. 50-276-4206 Supplier Grainger Supplier No. 16P074
The Grainger 16P074 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is UL/CSA approved. The 16P074 is ideal for driving pumps, fans, blowers, conveyors, and other industrial machinery requiring up to 3/4 horsepower.