Learn More
Grainger Plastic Strapping Hand Strapping 5/8
Catalog No. 502763744
Encompass_Preferred
The Grainger 16P049 Plastic Strapping Hand Strapping Tool is a lightweight hand-operated tensioning and sealing tool designed for applying and joining polypropylene strapping. It features a compact ergonomic design with rubber grip handles for comfort. This tool can apply up to 65 lbs of tension and will seal straps up to 5/8" wide and .020" thick using a heated element. It requires 110V power and has an 8 foot cord. The 16P049 is ideal for securing bundles, crates and skids in warehouses, shipping departments, and other industrial settings. This is a high-quality strapping tool made in the USA by Signode.
Catalog No. 50-276-3744 Supplier Grainger Supplier No. 16P049
The Grainger 16P049 Plastic Strapping Hand Strapping Tool is a lightweight hand-operated tensioning and sealing tool designed for applying and joining polypropylene strapping. It features a compact ergonomic design with rubber grip handles for comfort. This tool can apply up to 65 lbs of tension and will seal straps up to 5/8" wide and .020" thick using a heated element. It requires 110V power and has an 8 foot cord. The 16P049 is ideal for securing bundles, crates and skids in warehouses, shipping departments, and other industrial settings. This is a high-quality strapping tool made in the USA by Signode.