Learn More
Grainger Plastic Strapping Hand Strapping 1/2
Catalog No. 502764084
Encompass_Preferred
The 1-inch plastic strapping from Grainger is a general purpose polypropylene strapping designed for light to medium duty bundling, baling, and unitizing applications. It has a break strength of 55 lbs per inch width. The strapping comes in a coil with 16,000 feet of continuous length and 1 inch width. It is lightweight, flexible, weather and chemical resistant. This strapping works well for securing bundles of paper, cardboard, plastic, and wood products. It can be tensioned and sealed with a hand strapping tool.
Catalog No. 50-276-4084 Supplier Grainger Supplier No. 16P068
The 1-inch plastic strapping from Grainger is a general purpose polypropylene strapping designed for light to medium duty bundling, baling, and unitizing applications. It has a break strength of 55 lbs per inch width. The strapping comes in a coil with 16,000 feet of continuous length and 1 inch width. It is lightweight, flexible, weather and chemical resistant. This strapping works well for securing bundles of paper, cardboard, plastic, and wood products. It can be tensioned and sealed with a hand strapping tool.