Learn More
Grainger Plastic Strapping Hand Strapping 1/2
Catalog No. 502764484
Encompass_Preferred
The Grainger 16P066 plastic strapping is a general purpose polypropylene strapping that is used for unitizing and bundling items. It has a break strength of 525 lbs, a thickness of 0.020 inches, and comes in a coil with a length of 1500 feet. The strapping is blue in color. This plastic strapping can be used for a variety of applications including bundling boxes, bricks, paper, and more. It secures items with a friction weld.
Catalog No. 50-276-4484 Supplier Grainger Supplier No. 16P067
The Grainger 16P066 plastic strapping is a general purpose polypropylene strapping that is used for unitizing and bundling items. It has a break strength of 525 lbs, a thickness of 0.020 inches, and comes in a coil with a length of 1500 feet. The strapping is blue in color. This plastic strapping can be used for a variety of applications including bundling boxes, bricks, paper, and more. It secures items with a friction weld.