Learn More
Grainger Plastic Strapping Hand Strapping 3/4
Catalog No. 502763920
Encompass_Preferred
The Approved Vendor Plastic Strapping is a 2" wide polypropylene strapping that comes in a 500 ft roll. It has a break strength of 550 lbs and is recommended for general purpose bundling, palletizing, and unitizing applications. The strapping is lightweight but durable with good tensile strength. It can be used with both automatic and hand tools. This is an economical strapping choice for light to medium duty securing needs.
Catalog No. 50-276-3920 Supplier Grainger Supplier No. 16P071
The Approved Vendor Plastic Strapping is a 2" wide polypropylene strapping that comes in a 500 ft roll. It has a break strength of 550 lbs and is recommended for general purpose bundling, palletizing, and unitizing applications. The strapping is lightweight but durable with good tensile strength. It can be used with both automatic and hand tools. This is an economical strapping choice for light to medium duty securing needs.