Learn More
Grainger Plastic Strapping Kit Standard 1/2
Catalog No. 502764486
Encompass_Preferred
The Grainger Plastic Strapping Kit is a heavy duty plastic strapping kit for securing large items or bundles. It includes a dispenser, sealer and tensioner to apply 1-1/4" wide x .031" thick polypropylene strapping. The dispenser holds up to 500' of strapping. The sealer creates a strong seal using heat. The tensioner allows proper tensioning to secure the load. This kit provides an all in one solution for applying plastic strapping.
Catalog No. 50-276-4486 Supplier Grainger Supplier No. 16P047
The Grainger Plastic Strapping Kit is a heavy duty plastic strapping kit for securing large items or bundles. It includes a dispenser, sealer and tensioner to apply 1-1/4" wide x .031" thick polypropylene strapping. The dispenser holds up to 500' of strapping. The sealer creates a strong seal using heat. The tensioner allows proper tensioning to secure the load. This kit provides an all in one solution for applying plastic strapping.