Learn More
Grainger Plastic Strapping Machine Strapping 5/8
Catalog No. 502764603
Encompass_Preferred
The Signode 16P052 plastic strapping machine is a semi-automatic tabletop strapping machine designed for low to medium duty strapping applications. It features a 1/2 HP motor and can strap up to 60 bundles per hour. This polyester strapping machine has a maximum strap width of 5/8 inches and can apply tension up to 65 lbs. The 16P052 has an automatic heat seal mechanism with adjustable temperature control. This tabletop strapping machine measures 33 x 17 x 24 inches (LxWxH) and weighs approximately 100 lbs. Safety features include emergency stop button and lockout/tagout capability.
Catalog No. 50-276-4603 Supplier Grainger Supplier No. 16P052
The Signode 16P052 plastic strapping machine is a semi-automatic tabletop strapping machine designed for low to medium duty strapping applications. It features a 1/2 HP motor and can strap up to 60 bundles per hour. This polyester strapping machine has a maximum strap width of 5/8 inches and can apply tension up to 65 lbs. The 16P052 has an automatic heat seal mechanism with adjustable temperature control. This tabletop strapping machine measures 33 x 17 x 24 inches (LxWxH) and weighs approximately 100 lbs. Safety features include emergency stop button and lockout/tagout capability.