Learn More
Grainger Plastic Strapping Machine Strapping 5/8
Catalog No. 502764606
Encompass_Preferred
The Grainger Approved 1-1/2" x 0.020" x 1500' Plastic Strapping is a general purpose polypropylene strapping that is lightweight and economical. It has moderate tensile strength and is resistant to rot, mildew and moisture. This strapping is suitable for light- to medium-duty unitizing, bundling and palletizing applications. It comes in a dispenser box for convenient use. The strapping has a break strength of 55 lbs, meets military spec MIL-STD-105E, and is made in the USA.
Catalog No. 50-276-4606 Supplier Grainger Supplier No. 16P055
The Grainger Approved 1-1/2" x 0.020" x 1500' Plastic Strapping is a general purpose polypropylene strapping that is lightweight and economical. It has moderate tensile strength and is resistant to rot, mildew and moisture. This strapping is suitable for light- to medium-duty unitizing, bundling and palletizing applications. It comes in a dispenser box for convenient use. The strapping has a break strength of 55 lbs, meets military spec MIL-STD-105E, and is made in the USA.