Learn More
Research Products International Corp Plastic Syringes for Repetitive Dispensers, Non Sterile, 5ml Capacity, 100 per Case
SDP
Catalog No. 50999255
Encompass
Designed for exact fit with all leading manufacturers repetitive manual dispensers. Refillable syringes provide sufficient capacity to dispense up to different sample volumes. Syringes are made of high clarity polypropylene with a polyethylene plunger and feature printed graduations. Autoclaving is not recommended.
Catalog No. 50-999-255 Supplier Research Products International Corp Supplier No. 256106
Designed for exact fit with all leading manufacturers repetitive manual dispensers. Refillable syringes provide sufficient capacity to dispense up to different sample volumes. Syringes are made of high clarity polypropylene with a polyethylene plunger and feature printed graduations. Autoclaving is not recommended.