Learn More
Research Products International Corp Plastic Syringes, Sterile, 0.5ml, 100/cs
SDP
Supplier:  Research Products International Corp 256114
Encompass
Designed for exact fit with all leading manufacturers repetitive manual dispensers. Refillable syringes provide sufficient capacity to dispense up to different sample volumes. Syringes are made of high clarity polypropylene with a polyethylene plunger and feature printed graduations. Autoclaving is not recommended.
Catalog No. 50-197-7503