Learn More
Electron Microscopy Sciences Style 272, 1/EA
SDP
Catalog No. 502091080
Encompass_Preferred
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. Replaceable tip tweezers - straight, flat, squared. Length:5" (130mm). ESD safe.
Catalog No. 50-209-1080 Supplier Electron Microscopy Sciences Supplier No. 78325272
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. Replaceable tip tweezers - straight, flat, squared. Length:5" (130mm). ESD safe.