Learn More
Electron Microscopy Sciences Plastic Tissue Capsule
SDP
Catalog No. 5027881
Encompass_Preferred
Plastic tissue capsules molded from polyethylene and resistant to all processing chemicals, including Zenker's Fixative. A three-compartment divider which is removable for specimens up to 35 mm. A weighted end floats the capsule in a upright position to conserve space. Perforated on the top and bottom for better circulation of the fluid. Snap-on hinged cover.
Catalog No. 50-278-81 Supplier Electron Microscopy Sciences Supplier No. 6232212
Plastic tissue capsules molded from polyethylene and resistant to all processing chemicals, including Zenker's Fixative. A three-compartment divider which is removable for specimens up to 35 mm. A weighted end floats the capsule in a upright position to conserve space. Perforated on the top and bottom for better circulation of the fluid. Snap-on hinged cover.