Learn More
Ted Pella Inc Plastic Tweezer, Heat-Resistant, flat, 116mm, Ryton (PPS) 2.4 x 0.20mm
SDP
Catalog No. NC9308637
Encompass
For temperatures up to 220° C / 425° F when handling such items as microminiature components for soldering. Useful in chemical processing of semiconductors.
Catalog No. NC9308637 Supplier Ted Pella Inc Supplier No. 5616
For temperatures up to 220° C / 425° F when handling such items as microminiature components for soldering. Useful in chemical processing of semiconductors.