Learn More
Ted Pella Inc Tweezer, white, wafer style, 112mm, Fluorocarbon (CTFE), 2.16 x 0.18mm
SDP
Catalog No. NC9308631
Encompass
Useful for chemical processing. Resists hydrofluoric acid. Temperatures up to 400°F / 204°C.
Catalog No. NC9308631 Supplier Ted Pella Inc Supplier No. 53011
Useful for chemical processing. Resists hydrofluoric acid. Temperatures up to 400°F / 204°C.