Learn More
Fine Science Tools Plastic Caliper, 0.1 mm Increments, 190 mm Max Measurement
SDP
Catalog No. NC9280794
Encompass
Product: Plastic Caliper/ Alloy/Material: Plastic/ Increments: 0.1mm/ Max Measurement: 190mm/ Features: Tough polymer is unbreakable under normal conditions. Measures inside, outside, step and depth/ Total Length: 21 cm/ Unit: Each
Catalog No. NC9280794 Supplier Fine Science Tools Supplier No. 3008720
Product: Plastic Caliper/ Alloy/Material: Plastic/ Increments: 0.1mm/ Max Measurement: 190mm/ Features: Tough polymer is unbreakable under normal conditions. Measures inside, outside, step and depth/ Total Length: 21 cm/ Unit: Each