Learn More
Electronic Imaging Materials Inc 667 Vial CryoLabel 1.00 inch x 0.40 inch, White, 1 inch core, 4.5K/roll
SDP
Catalog No. NC1229608
Encompass_Preferred
1.00 inch x 0.40 inch white vial CryoLabel handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC1229608 Supplier Electronic Imaging Materials Inc Supplier No. 92260
1.00 inch x 0.40 inch white vial CryoLabel handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.