Learn More
Research Products International Corp Plastic Weighing Dishes, Natural, 1 5/8x5/16"", 4000/pk
SDP
Catalog No. 501977705
Encompass
Weighing Dishes will resist diluted acids, aqueous solutions, alcohols and bases. They are ideal for many applications such as weighing, dispensing or storing. They are safe, contaminant-free, biologically inert economical containers for weighing liquid or powdered samples in the laboratory. Flat bottom ensures perfect stability on countertops. They have a smooth surface providing accurate pour-outs with minimal sample loss and facilitating weighing of static-affected samples. Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Will withstand temperatures up to 80ºC.
This item is not returnable. View return policy
Catalog No. 50-197-7705 Supplier Research Products International Corp Supplier No. 142159
Weighing Dishes will resist diluted acids, aqueous solutions, alcohols and bases. They are ideal for many applications such as weighing, dispensing or storing. They are safe, contaminant-free, biologically inert economical containers for weighing liquid or powdered samples in the laboratory. Flat bottom ensures perfect stability on countertops. They have a smooth surface providing accurate pour-outs with minimal sample loss and facilitating weighing of static-affected samples. Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Will withstand temperatures up to 80ºC.