Learn More
Chemglass Life Sciences Plastic Lab Safety Razor, Large
SDP
Supplier:  Chemglass Life Sciences CLS191102
Encompass_Preferred
Plastic Lab Safety Razor, Large. Made in the USA. At just 0.27mm (0.011) the US Patent Pending smartSlicer is as thin as a single edge industrial razor blade but much safer and unlikely to slice you! The smartSlicer is able to cleanly slice gels* right on the light Box without scratching the surface of the box. The smartSlicer can also be used to chop, mince or slice some soft tissue samples. The smartSlicer is also adjustable - simply cut it with standard scissors to the width you require. Large: 50mm L x 25mm H x 0.27mm Thick.
Catalog No. 50-153-2254