Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled Precision Pump Tubing, Platinum-Cured Silicone, L/S 25; 500 ft
Catalog No. NC9923475
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC9923475 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640325/DEL
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.