Learn More
Cole-Parmer Platinum-Cured Silicone Tubing, 50 Shore A, 1/16" ID x 3/16" OD; 50 Ft
Catalog No. 502563461 Shop All Cole-Parmer Products
Encompass_Preferred
Produced with very high dimensional tolerances, the platinum-cured silicone tubing provides the accuracy and purity needed for critical biopharmaceutical applications. This soft yet durable tubing offers a long service life in peristaltic pump or pinch valve use.
Easily maintain the purity of the fluid—tubing does not contain BPA, phthalates, or peroxides that can leach out into the fluid. Tubing is manufactured, double-bagged, and sealed in an ISO Class 7 cleanroom. For an added level of particle control, a liner is placed between the double-bagged tubing package and the box.
Catalog No. 50-256-3461 Supplier Cole-Parmer Supplier No. 5011964
Produced with very high dimensional tolerances, the platinum-cured silicone tubing provides the accuracy and purity needed for critical biopharmaceutical applications. This soft yet durable tubing offers a long service life in peristaltic pump or pinch valve use.
Easily maintain the purity of the fluid—tubing does not contain BPA, phthalates, or peroxides that can leach out into the fluid. Tubing is manufactured, double-bagged, and sealed in an ISO Class 7 cleanroom. For an added level of particle control, a liner is placed between the double-bagged tubing package and the box.