Learn More
Thomas Scientific Deep Well Plates - 384 Well, Volume: 200æl, Height: 22mm, Qty: 24
SDP
Catalog No. 501458610
Encompass
100µl, 200µl, 300µl, 400µl well volume Standard dimensions / robot compatible Polypropylene 384 well plates are designed for HTS applications and are compatible with multichannel pipettes and robotic workstations. All sizes are standard 127.8 x 85.5mm dimensions. All sizes have rounded well bottoms except B9384-1.
Catalog No. 50-145-8610 Supplier Thomas Scientific Supplier No. 1159N33
100µl, 200µl, 300µl, 400µl well volume Standard dimensions / robot compatible Polypropylene 384 well plates are designed for HTS applications and are compatible with multichannel pipettes and robotic workstations. All sizes are standard 127.8 x 85.5mm dimensions. All sizes have rounded well bottoms except B9384-1.