Learn More
Molecular Devices Molecular Devices Petri Dish Holder, 1-Way Adjustable, 138-141 mm, Spring-Loaded
Catalog No. 5000014498
Encompass_Preferred
Molecular Devices' adjustable petri dish and microplate holder secures plastic plates and dishes for automated colony-picking and imaging workflows. The spring-loaded, one-way adjustable design accommodates plate diameter variation while minimizing light scatter with a black, low-reflectance finish.
  • One-way spring-loaded adjustment for secure plate retention.
  • Compatible with plates 138-141 mm in diameter.
  • Black low-reflectance finish reduces light scattering for improved imaging.
  • Fits common automated colony-picking and microplate handling systems.
  • Durable construction for repeated use in laboratory workflows.
Catalog No. 50-000-14498 Supplier Molecular Devices Supplier No. X9401
Molecular Devices' adjustable petri dish and microplate holder secures plastic plates and dishes for automated colony-picking and imaging workflows. The spring-loaded, one-way adjustable design accommodates plate diameter variation while minimizing light scatter with a black, low-reflectance finish.
  • One-way spring-loaded adjustment for secure plate retention.
  • Compatible with plates 138-141 mm in diameter.
  • Black low-reflectance finish reduces light scattering for improved imaging.
  • Fits common automated colony-picking and microplate handling systems.
  • Durable construction for repeated use in laboratory workflows.