Learn More
MTC Bio Plate 96 Round Well V-Bottom 14 40mm Height 480uL Non-Sterile 10 Plates/Pack 10 Packs/Case
Catalog No. 5000460086
Encompass_Preferred
Plate 96 Round Well V-Bottom 14 40mm Height 480uL Non-Sterile 10 Plates/Pack 10 Packs/Case
Catalog No. 50-004-60086 Supplier MTC Bio Supplier No. DP33VR9N
Plate 96 Round Well V-Bottom 14 40mm Height 480uL Non-Sterile 10 Plates/Pack 10 Packs/Case