Learn More
Meso Scale Discovery MSD GOLD 96-well Streptavidin SECTOR Plate
SDP
Supplier:  Meso Scale Discovery L15SA1
Encompass
MSD GOLD Streptavidin plates bind biotin-conjugated molecules for flexible and rapid assay development. These streptavidin-coated plates are designed to be the gold standard for assay performance, consistency, and convenience. MSD GOLD Streptavidin plates are highly suited for use with homogenous assays or bridging assays, such as typical immunogenicity assays that require high free drug tolerance. They are also ideal for pharmacokinetic / pharmacodynamic studies, and biomarker assay development, and screening applications. Every MSD GOLD Streptavidin plate lot meets consistent release criteria specified on the certificate of analysis and has an extended shelf life of 30 months, making them ideal for longitudinal studies.
Catalog No. 50-914-703