Learn More
Andwin Scientific PLATE 96 WELL FLAT BTM
SDP
Catalog No. NC9181191
Encompass_Preferred
The 96-Well Plate is constructed with polystyrene. This product has a flat bottom with a chimney style back. It does not have a lit and it has a medium binding. The plate is also non-sterile.
Catalog No. NC9181191 Supplier Andwin Scientific Supplier No. 655096
Add to Cart
Add to Cart
The 96-Well Plate is constructed with polystyrene. This product has a flat bottom with a chimney style back. It does not have a lit and it has a medium binding. The plate is also non-sterile.