Learn More
Thomas Scientific Krystal 96 Well Microplate, Black, tissue culture treated with lid, individually packed, Bottom Flat, 350æl Well Capacity, Qty: 100
SDP
Catalog No. 501460876
Encompass
Opaque, Clear Bottom 96 Well Microplates These white and black plates are useful for applications such as fluorescence, luminescence and scintillation in instruments reading from below or above the wells. Designed to SBS specifications.
Catalog No. 50-146-0876 Supplier Thomas Scientific Supplier No. 1158A49
Opaque, Clear Bottom 96 Well Microplates These white and black plates are useful for applications such as fluorescence, luminescence and scintillation in instruments reading from below or above the wells. Designed to SBS specifications.