Learn More
Thomas Scientific Culture Plate, Flat, Low Evap. Lid
SDP
Catalog No. 501458935
Encompass
Compatible with most automatic dilutors, pipettors and centrifuges, plate readers and washers Alphanumeric markings aid in well identification Stacking beads and uniform footprints for easier handling Uniform footprint Costar 96 WellCell Culture Clusters are treated for optimal cell attachment and sterilized by gamma radiation. Culture clusters are certified non-pyrogenic. Most come with one-way lid and individual condensation rings. Packaged individually in cases of 50 or 100 or as noted. 6906A01 and A05 are fitted with special low evaporation lids. 6906A15 is a non-treated half area plate with reduced well diameters but the same 9 mm well-to-well spacing and half the growth area of the regular plates (0.16 cm 2 veresus 0.32 cm 2 ). 6906A21 has slightly more growth area (0.38 cm 2 ). 6906A23 are 8 well strips assembled 12 strips per plate.
This item is not returnable. View return policy
Catalog No. 50-145-8935 Supplier Thomas Scientific Supplier No. 6906A01
Compatible with most automatic dilutors, pipettors and centrifuges, plate readers and washers Alphanumeric markings aid in well identification Stacking beads and uniform footprints for easier handling Uniform footprint Costar 96 WellCell Culture Clusters are treated for optimal cell attachment and sterilized by gamma radiation. Culture clusters are certified non-pyrogenic. Most come with one-way lid and individual condensation rings. Packaged individually in cases of 50 or 100 or as noted. 6906A01 and A05 are fitted with special low evaporation lids. 6906A15 is a non-treated half area plate with reduced well diameters but the same 9 mm well-to-well spacing and half the growth area of the regular plates (0.16 cm 2 veresus 0.32 cm 2 ). 6906A21 has slightly more growth area (0.38 cm 2 ). 6906A23 are 8 well strips assembled 12 strips per plate.