Learn More
Thomas Scientific 96 Well Solid Microplates, Polystyrene, clear, Flat well bottom, 325æl Capacity, Inner Pack: 10, 100/PK
SDP
Catalog No. 501460871
Encompass
350ul Well Volume These 96 well solid microplates are available in black, white, and clear polystyrene or natural polypropylene. The colored plates are ideal for fluorescence, luminescence and scintillation while the clear plates are useful for ELISA assays. Designed to SBS specifications.
Catalog No. 50-146-0871 Supplier Thomas Scientific Supplier No. 1158A43
350ul Well Volume These 96 well solid microplates are available in black, white, and clear polystyrene or natural polypropylene. The colored plates are ideal for fluorescence, luminescence and scintillation while the clear plates are useful for ELISA assays. Designed to SBS specifications.