Learn More
Grainger Guardian Fall Protection 4-Way Plate Anchor, Steel, 1 ea.
Catalog No. 502795207
Encompass_Preferred
The 4-Way Plate Anchor is a versatile, low-profile fall protection anchor designed for use on wood, metal, or concrete substrates. Engineered for durability, this reusable anchor is suitable for pan decking or form work and serves as an effective endpoint for single or multi-span horizontal lifeline systems.
  • Supports one worker in fall arrest or two workers in fall restraint
  • Constructed from durable powder-coated steel
  • Compatible with metal substrates as thin as 24 ga
  • Meets OSHA 1910.140, OSHA 1926.502, and ANSI Z359.1–2007 standards
  • Minimum breaking strength of 5,000 lbs.
  • Worker capacity range of 130 lbs. to 420 lbs.
Catalog No. 50-279-5207 Supplier Grainger Supplier No. 19UC74
The 4-Way Plate Anchor is a versatile, low-profile fall protection anchor designed for use on wood, metal, or concrete substrates. Engineered for durability, this reusable anchor is suitable for pan decking or form work and serves as an effective endpoint for single or multi-span horizontal lifeline systems.
  • Supports one worker in fall arrest or two workers in fall restraint
  • Constructed from durable powder-coated steel
  • Compatible with metal substrates as thin as 24 ga
  • Meets OSHA 1910.140, OSHA 1926.502, and ANSI Z359.1–2007 standards
  • Minimum breaking strength of 5,000 lbs.
  • Worker capacity range of 130 lbs. to 420 lbs.