Learn More
Med Vet International Plate Bending Pliers, 15in, J0852
Catalog No. 5000584751
Encompass_Preferred
Double-sided roller-type. Used for 3.5mm narrow, 3.5mm broad and 4.5mm narrow plates. Stainless steel with aluminum handles.
Catalog No. 50-005-84751 Supplier Med Vet International Supplier No. JOR852
Double-sided roller-type. Used for 3.5mm narrow, 3.5mm broad and 4.5mm narrow plates. Stainless steel with aluminum handles.