Learn More
Grainger Pro-Line Plate Caster Kit, 4 in Wheel, Polyurethane, 1,000 lb Load Capacity, 4-Piece Set
Catalog No. 502791747
Encompass_Preferred
This plate caster kit is designed for DN series modular workstations, providing mobility for heavy-duty applications. The kit includes four casters, consisting of two swivel units with brakes and two rigid units. These casters feature a durable polyurethane wheel construction and a zinc-plated steel frame, ensuring robust performance and non-marking operation in various environments. This factory-installed option adds 3.25 in. to the overall height of the workbench.
  • Includes two swivel casters with total lock brakes and two rigid casters
  • Features non-marking polyurethane wheels with precision ball bearings
  • Utilizes a durable zinc-plated steel frame
  • Provides an overall load capacity of 1,000 lb.
  • Operates within a temperature range of -40°F to +180°F
  • Compatible with DN series adjustable height workstations
  • Includes mounting hardware for installation
Catalog No. 50-279-1747 Supplier Grainger Supplier No. 16D644
This plate caster kit is designed for DN series modular workstations, providing mobility for heavy-duty applications. The kit includes four casters, consisting of two swivel units with brakes and two rigid units. These casters feature a durable polyurethane wheel construction and a zinc-plated steel frame, ensuring robust performance and non-marking operation in various environments. This factory-installed option adds 3.25 in. to the overall height of the workbench.
  • Includes two swivel casters with total lock brakes and two rigid casters
  • Features non-marking polyurethane wheels with precision ball bearings
  • Utilizes a durable zinc-plated steel frame
  • Provides an overall load capacity of 1,000 lb.
  • Operates within a temperature range of -40°F to +180°F
  • Compatible with DN series adjustable height workstations
  • Includes mounting hardware for installation