Learn More
Grainger PRO-LINE Plate Caster Kit, Polyurethane, 4 in, Swivel Locking, Pkg. of 4
Catalog No. 502792059
Encompass_Preferred
This plate caster kit is designed for workbenches, allowing units to be easily repositioned or moved into different work areas. These heavy-duty casters feature a total lock brake system that secures both the swivel and the wheel simultaneously for enhanced stability.
  • Includes four swivel locking casters
  • Total lock brake mechanism secures wheel and swivel
  • Equipped with double ball bearing raceway
  • Durable polyurethane wheel material for non-marking operation
  • Compatible with series BT, HD, HDL, and MVSII workbenches
  • ESD safe construction
  • Includes necessary mounting hardware
  • Supports a total load capacity of 1,000 lb
  • Operating temperature range of -40°F to +180°F
Catalog No. 50-279-2059 Supplier Grainger Supplier No. 16D641
This plate caster kit is designed for workbenches, allowing units to be easily repositioned or moved into different work areas. These heavy-duty casters feature a total lock brake system that secures both the swivel and the wheel simultaneously for enhanced stability.
  • Includes four swivel locking casters
  • Total lock brake mechanism secures wheel and swivel
  • Equipped with double ball bearing raceway
  • Durable polyurethane wheel material for non-marking operation
  • Compatible with series BT, HD, HDL, and MVSII workbenches
  • ESD safe construction
  • Includes necessary mounting hardware
  • Supports a total load capacity of 1,000 lb
  • Operating temperature range of -40°F to +180°F