Learn More
Grainger Plate Caster Kit Polyurethane 275 lb PK4
Catalog No. 502768120
Encompass_Preferred
The Grainger Approved Plate Caster Kit includes 4 swivel casters with a 3" diameter, 1-1/4" wide plate, and a 125 lb weight capacity per caster. The casters feature non-marking gray rubber wheels and zinc-plated steel forks, plates, stems, and nuts. The stem of each caster is 3-1/2" long and can be mounted with the included nuts and washers. This caster kit provides a smooth-rolling, heavy duty swiveling option for equipment mobility.
Catalog No. 50-276-8120 Supplier Grainger Supplier No. 16Y922
The Grainger Approved Plate Caster Kit includes 4 swivel casters with a 3" diameter, 1-1/4" wide plate, and a 125 lb weight capacity per caster. The casters feature non-marking gray rubber wheels and zinc-plated steel forks, plates, stems, and nuts. The stem of each caster is 3-1/2" long and can be mounted with the included nuts and washers. This caster kit provides a smooth-rolling, heavy duty swiveling option for equipment mobility.